Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CANX SIRNA Assays ›
          Gene Symbol:
          Canx
          Gene Name:
          calnexin
          Gene Aliases:
          1110069N15Rik, AI988026, Cnx, D11Ertd153e
          Chromosome Location:
          Chr.11: 50293957 - 50325673 on Build GRCm38
          UniGene ID:
          Mm.248827
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 159972

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Canx
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001110499.1 NP_001103969.1 9 1170
          NM_001110500.1 NP_001103970.1 9 1138
          NM_007597.3 NP_031623.1 9 1174
          GenBank mRNA AK028279.1 - 9 1102
          AK084175.1 - 9 1114
          AK159298.1 - 9 1158
          AK165508.1 - 9 1073
          AK166649.1 - 9 1151
          BC012408.1 - 9 1092 3497769
          BC040244.1 - 9 1101 2582119
          L18888.1 - 8 986

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          synaptic vesicle endocytosis
          chaperone-mediated protein folding
          clathrin-mediated endocytosis
          glycoprotein binding
          calcium ion binding
          protein binding
          carbohydrate binding
          apolipoprotein binding
          ionotropic glutamate receptor binding
          poly(A) RNA binding
          metal ion binding
          unfolded protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qvstf:80/100.66.73.196:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0