Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007619.2 | NP_031645.2 | 7 | 1204 | ||
GenBank mRNA | AK029826.1 | - | 7 | 1207 | ||
AK153915.1 | - | 7 | 1156 | |||
AK163907.1 | - | 7 | 1160 | |||
BC125285.1 | - | 7 | 1119 | 40129800 | ||
X57111.1 | - | 7 | 1142 |