Search
Search

Ccnf| Gene Symbol: Ccnf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007634.4 | NP_031660.3 | 5 | 627 | ||
| GenBank mRNA | AK030298.1 | - | 566 | |||
| AK031613.1 | - | 5 | 567 | |||
| AK032961.1 | - | 5 | 553 | |||
| AK082485.1 | - | 5 | 595 | |||
| AK085183.1 | - | 5 | 564 | |||
| AK159594.1 | - | 5 | 628 | |||
| AK162447.1 | - | 5 | 567 | |||
| AK169263.1 | - | 5 | 578 | |||
| BC021658.1 | - | 5 | 547 | |||
| BC037662.1 | - | 5 | 579 | 5353169 | ||
| U20612.1 | - | 5 | 565 | |||
| U20636.1 | - | 5 | 682 | |||
| Z47766.1 | - | 5 | 567 | |||