Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDKN1A SIRNA Assays ›
          Gene Symbol:
          Cdkn1a 4930567H12Rik
          Gene Name:
          cyclin-dependent kinase inhibitor 1A (P21)
          RIKEN cDNA 4930567H12 gene
          Gene Aliases:
          CAP20, CDKI, CIP1, Cdkn1, P21, SDI1, Waf1, mda6, p21Cip1, p21WAF
          4930579P07Rik
          Chromosome Location:
          Chr.17: 29090986 - 29100722 on Build GRCm38
          Chromosome Location:
          Chr.8: 125584638 - 125616670 on Build GRCm38
          UniGene ID:
          Mm.195663
          Mm.195663
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 160140

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cdkn1a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001111099.1 NP_001104569.1 2, 3 554
          NM_007669.4 NP_031695.1 2, 3 528
          GenBank mRNA AB017817.1 - 1, 2 427
          AB017818.1 - 1, 2 427
          AK007630.1 - 2, 3 526
          AK089093.1 - 2, 3 526
          AK139306.1 - 2, 3 529
          AK149910.1 - 2, 3 529
          AK170467.1 - 2, 3 529
          AK194916.1 - 1, 2 41
          AK210703.1 - 12
          BC002043.1 - 2, 3 481 3495942
          CT010218.1 - 1, 2 427
          U09507.1 - 1, 2 627
          U24173.1 - 2, 3 519
          Gene Symbol: 4930567H12Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK131763.1 - 810

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cyclin-dependent protein serine/threonine kinase activity
          G1/S transition of mitotic cell cycle
          G2/M transition of mitotic cell cycle
          protein phosphorylation
          cellular response to DNA damage stimulus
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          cell cycle
          cell cycle arrest
          regulation of mitotic cell cycle
          negative regulation of cell proliferation
          response to UV
          negative regulation of gene expression
          negative regulation of cell growth
          positive regulation of B cell proliferation
          cellular response to extracellular stimulus
          regulation of protein import into nucleus, translocation
          negative regulation of phosphorylation
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          negative regulation of apoptotic process
          positive regulation of programmed cell death
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of protein kinase activity
          positive regulation of fibroblast proliferation
          regulation of cell cycle
          cellular response to ionizing radiation
          cellular response to UV-B
          mitotic cell cycle arrest
          signal transduction by p53 class mediator
          cellular senescence
          negative regulation of G1/S transition of mitotic cell cycle
          regulation of DNA biosynthetic process
          positive regulation of reactive oxygen species metabolic process
          protein kinase inhibitor activity
          cyclin-dependent protein serine/threonine kinase inhibitor activity
          protein binding
          cyclin-dependent protein kinase activating kinase activity
          cyclin binding
          ubiquitin protein ligase binding
          protein complex binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0