Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF033566.1 | - | 12 | 1305 | ||
AK013974.2 | - | 10 | 1060 | |||
AY653308.1 | - | 11 | 1195 | |||
BC002220.1 | - | 8 | 861 | 3490616 | ||
BC012675.1 | - | 12 | 1327 | 3982920 | ||
CT010293.1 | - | 11 | 1227 |