Search
Search

Mapre1| Gene Symbol: Mapre1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007896.3 | NP_031922.1 | 7 | 965 | ||
| GenBank mRNA | AK013626.1 | - | 7 | 905 | ||
| AK146206.1 | - | 7 | 883 | |||
| AK154245.1 | - | 7 | 909 | |||
| AK179230.1 | - | 1 | 64 | |||
| BC043333.1 | - | 7 | 929 | |||
| BC052405.1 | - | 6 | 840 | |||
| BC052728.1 | - | 876 | ||||
| BC064444.1 | - | 876 | 6815724 | |||
| U51196.1 | - | 7 | 895 | |||