Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK010261.1 | - | 4 | 890 | ||
AK019319.1 | - | 1 | 625 | |||
AK131624.1 | - | 4 | 890 | |||
AK134921.1 | - | 4 | 890 | |||
AK149568.1 | - | 4 | 872 | |||
AK150834.1 | - | 4 | 892 | |||
AK159105.1 | - | 4 | 890 | |||
AK159424.1 | - | 4 | 892 | |||
AK159517.1 | - | 4 | 890 | |||
AK197774.1 | - | 1 | 17 | |||
BC028816.1 | - | 4 | 869 | 5136415 | ||
BC044785.1 | - | 3 | 861 | |||
BC083351.1 | - | 3 | 859 | 6818357 | ||
CT010212.1 | - | 3 | 824 | |||
CT010356.1 | - | 3 | 824 | |||
M12414.1 | - | 3 | 824 |