Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290788.1 | NP_001277717.1 | 7, 8 | 718 | ||
NM_009702.3 | NP_033832.2 | 7, 8 | 718 | |||
GenBank mRNA | AK016322.1 | - | 7, 8 | 717 | ||
AK077886.1 | - | 7, 8 | 708 | |||
AK082873.1 | - | 7, 8 | 657 | |||
AK122310.1 | - | 7, 8 | 706 | |||
AK138700.1 | - | 7, 8 | 774 | |||
AK145547.1 | - | 7, 8 | 703 | |||
AK151716.1 | - | 7, 8 | 689 | |||
AK202335.1 | - | 2, 3 | 160 | |||
BC042479.1 | - | 7, 8 | 698 | 5363276 | ||
U90333.1 | - | 7, 8 | 566 |