Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110140.3 | NP_001103610.1 | 1, 2 | 665 | ||
NM_009722.3 | NP_033852.1 | 1, 2 | 665 | |||
GenBank mRNA | AJ131821.1 | - | 1, 2 | 335 | ||
AJ223584.1 | - | 1, 2 | 335 | |||
AK220538.1 | - | 1, 2 | 614 | |||
BC054531.1 | - | 1, 2 | 324 | 6406916 | ||
BC054748.1 | - | 1, 2 | 324 | 6406916 |