Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009740.2 | NP_033870.1 | 2 | 395 | ||
GenBank mRNA | AF057701.1 | - | 2 | 279 | ||
AF100339.1 | - | 2 | 287 | |||
AF127387.1 | - | 2 | 198 | |||
AF134396.1 | - | 2 | 279 | |||
AJ006289.1 | - | 2 | 276 | |||
AK076082.1 | - | 2 | 307 | |||
AK140179.1 | - | 2 | 267 | |||
AK146236.1 | - | 2 | 301 | |||
AK150883.1 | - | 2 | 296 | |||
AK152563.1 | - | 2 | 296 | |||
AK152847.1 | - | 2 | 292 | |||
AK156890.1 | - | 2 | 349 | |||
AK168084.1 | - | 2 | 301 | |||
AK169126.1 | - | 2 | 302 | |||
AK172158.1 | - | 2 | 306 | |||
BC003703.1 | - | 2 | 314 | |||
BC024379.1 | - | 2 | 273 | 4976147 | ||
BC172007.1 | - | 2 | 279 | 9054293 | ||
BC172009.1 | - | 2 | 279 | 9054295 | ||
BU707261.1 | - | 2 | 395 |