Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Bub3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009774.3 | NP_033904.2 | 3 | 318 | ||
GenBank mRNA | AF149822.1 | - | 2 | 254 | ||
AK088534.1 | - | 3 | 319 | |||
AK190470.1 | - | 2 | 120 | |||
BC025089.1 | - | 3 | 330 | 3586753 | ||
BC086790.1 | - | 2 | 557 | |||
U67327.1 | - | 3 | 305 |