Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC145905.1 | - | 3, 4 | 455 | ||
BC145911.1 | - | 3, 4 | 455 | |||
L29221.1 | - | 3, 4 | 468 | |||
U21209.1 | - | 3, 4 | 468 |