Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CXCR2 SIRNA Assays ›
Gene Symbol:
Cxcr2
Gene Name:
chemokine (C-X-C motif) receptor 2
Gene Aliases:
CD128, CDw128, Cmkar2, Gpcr16, IL-8Rh, IL-8rb, IL8RA, Il8rb, mIL-8RH
Chromosome Location:
Chr.1: 74153994 - 74161246 on Build GRCm38
UniGene ID:
Mm.234466
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
161229
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Cxcr2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_009909.3
NP_034039.1
2
1283
GenBank mRNA
AK158234.1
-
3
1289
BC051677.1
-
3
1256
30023687
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008767215, XM_008767214, XM_008767213.
Set Membership:
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
acute inflammatory response to antigenic stimulus
positive regulation of leukocyte chemotaxis
chemotaxis
cellular defense response
signal transduction
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
positive regulation of cell proliferation
positive regulation of cardiac muscle cell apoptotic process
neutrophil chemotaxis
receptor internalization
negative regulation of neutrophil apoptotic process
interleukin-8-mediated signaling pathway
neutrophil activation
negative regulation of apoptotic process
positive regulation of vascular permeability
positive regulation of angiogenesis
chemokine-mediated signaling pathway
metanephric tubule morphogenesis
positive regulation of neutrophil chemotaxis
signal transducer activity
interleukin-8 receptor activity
G-protein coupled receptor activity
chemokine receptor activity
protein binding
C-X-C chemokine receptor activity
interleukin-8 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0