Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CRHR2 SIRNA Assays ›
          Gene Symbol:
          Crhr2
          Gene Name:
          corticotropin releasing hormone receptor 2
          Gene Aliases:
          CRF-R2, CRFR2alpha, CRFR2beta, Crfr2
          Chromosome Location:
          Chr.6: 55090048 - 55133016 on Build GRCm38
          UniGene ID:
          Mm.236081
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 161326

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Crhr2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001288618.1 NP_001275547.1 10 1139
          NM_001288619.1 NP_001275548.1 9 1046
          NM_001288620.1 NP_001275549.1 8 936
          NM_009953.4 NP_034083.2 10 1136
          GenBank mRNA AK142261.1 - 10 1086
          AY445512.1 - 9 1046
          AY753668.1 - 8 1008
          BC137592.1 - 10 1022 8860611
          FJ548565.1 - 10 964
          U17858.1 - 10 964
          U19939.1 - 10 1092
          U21729.1 - 10 1090

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          actin filament organization
          signal transduction
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway
          protein kinase C-activating G-protein coupled receptor signaling pathway
          hormone-mediated signaling pathway
          positive regulation of heart rate
          positive regulation of gene expression
          negative regulation of gene expression
          negative regulation of norepinephrine secretion
          positive regulation of serotonin secretion
          negative regulation of angiogenesis
          sensory perception of pain
          positive regulation of cAMP metabolic process
          negative regulation of cAMP biosynthetic process
          positive regulation of cAMP biosynthetic process
          epithelial cell differentiation
          positive regulation of interleukin-6 production
          negative regulation of epinephrine secretion
          positive regulation of stress-activated MAPK cascade
          negative regulation of luteinizing hormone secretion
          gastric motility
          catecholamine biosynthetic process
          positive regulation of cAMP-mediated signaling
          negative regulation of cAMP-mediated signaling
          positive regulation of blood pressure
          negative regulation of follicle-stimulating hormone secretion
          skeletal muscle tissue growth
          positive regulation of NF-kappaB transcription factor activity
          long-term synaptic potentiation
          negative regulation of insulin secretion involved in cellular response to glucose stimulus
          regulation of ERK1 and ERK2 cascade
          cellular response to corticotropin-releasing hormone stimulus
          negative regulation of calcium ion import
          negative regulation of feeding behavior
          negative regulation of defecation
          signal transducer activity
          transmembrane signaling receptor activity
          G-protein coupled receptor activity
          hormone activity
          protein binding
          corticotrophin-releasing factor receptor activity
          peptide hormone binding
          corticotropin-releasing hormone receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0