Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289813.1 | NP_001276742.1 | 2 | 271 | ||
NM_001289814.1 | NP_001276743.1 | 2 | 271 | |||
NM_010047.4 | NP_034177.1 | 3 | 468 | |||
GenBank mRNA | AF021031.1 | - | 2 | 172 | ||
AK019346.1 | - | 3 | 471 | |||
AK160342.1 | - | 3 | 458 | |||
BC048503.1 | - | 2 | 123 | 6770478 | ||
BE945982.1 | - | 2 | 171 |