Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146308.1 | NP_001139780.1 | 1, 2 | 162 | ||
NM_001146309.1 | NP_001139781.1 | 1, 2 | 162 | |||
NM_013810.3 | NP_038838.1 | 1, 2 | 162 | |||
GenBank mRNA | AK046073.1 | - | 1, 2 | 142 | ||
AK053796.1 | - | 1, 2 | 158 | |||
AK078082.1 | - | 1, 2 | 129 | |||
AK134136.1 | - | 1, 2 | 129 | |||
AK134487.1 | - | 1, 2 | 129 | |||
AK140752.1 | - | 1, 2 | 129 | |||
AK142818.1 | - | 1, 2 | 112 | |||
AK146920.1 | - | 1, 2 | 129 | |||
AK151096.1 | - | 1, 2 | 142 | |||
AK152110.1 | - | 1, 2 | 129 | |||
AK152204.1 | - | 1, 2 | 142 | |||
AK153072.1 | - | 1, 2 | 142 | |||
AK153389.1 | - | 1, 2 | 116 | |||
AK159984.1 | - | 1, 2 | 139 | |||
AK161973.1 | - | 1, 2 | 142 | |||
AK167956.1 | - | 1, 2 | 102 | |||
AK172471.1 | - | 1, 2 | 119 | |||
AK199448.1 | - | 1, 2 | 39 | |||
AY098595.1 | - | 1, 2 | 85 | |||
BC046430.1 | - | 1, 2 | 127 | 4986658 | ||
BY293844.1 | - | 1, 2 | 162 | |||
U58884.1 | - | 1, 2 | 115 |