Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000142.4 | NP_000133.1 | 18 | 3264 | ||
NM_001163213.1 | NP_001156685.1 | 18 | 3270 | |||
NM_022965.3 | NP_075254.1 | 16 | 2928 | |||
XM_006713868.1 | XP_006713931.1 | 18 | 3292 | |||
XM_006713869.1 | XP_006713932.1 | 18 | 3289 | |||
XM_006713870.1 | XP_006713933.1 | 18 | 3289 | |||
XM_006713871.1 | XP_006713934.1 | 18 | 3286 | |||
XM_006713872.1 | XP_006713935.1 | 18 | 3280 | |||
XM_006713873.1 | XP_006713936.1 | 18 | 3280 | |||
XM_011513420.1 | XP_011511722.1 | 18 | 3286 | |||
XM_011513422.1 | XP_011511724.1 | 18 | 3283 | |||
GenBank mRNA | AB209441.1 | - | 17 | 3199 | ||
M64347.1 | - | 16 | 2778 |