Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019734.2 | NP_062708.1 | 14 | 1233 | ||
GenBank mRNA | AF157500.1 | - | 14 | 1163 | ||
AF352179.1 | - | 14 | 1175 | |||
AK034204.1 | - | 1623 | ||||
AK075666.1 | - | 14 | 1148 | |||
AK075919.1 | - | 14 | 1187 | |||
AK077721.1 | - | 14 | 1186 | |||
AK090314.1 | - | 14 | 1183 | |||
AK150567.1 | - | 14 | 1145 | |||
AK151208.1 | - | 14 | 1234 | |||
AK151312.1 | - | 14 | 1145 | |||
AK151478.1 | - | 14 | 1224 | |||
AK152305.1 | - | 14 | 1185 | |||
AK152811.1 | - | 14 | 1146 | |||
AK152920.1 | - | 14 | 1194 | |||
AK153101.1 | - | 14 | 1194 | |||
AK153297.1 | - | 14 | 1147 | |||
AK159556.1 | - | 14 | 1155 | |||
AK166757.1 | - | 14 | 1165 | |||
BC003204.1 | - | 14 | 1153 | 3587042 |