Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161456.1 | NP_001154928.1 | 3 | 487 | ||
NM_001161457.1 | NP_001154929.1 | 4 | 604 | |||
NM_001161458.1 | NP_001154930.1 | 4 | 604 | |||
NM_022309.4 | NP_071704.3 | 4 | 604 | |||
GenBank mRNA | AK089094.1 | - | 4 | 605 | ||
AK134449.1 | - | 3 | 172 | |||
AK198845.1 | - | 54 | ||||
BC006763.1 | - | 4 | 372 | 3481800 | ||
BC026749.1 | - | 4 | 414 | 4016060 | ||
BC040752.1 | - | 4 | 403 | 4016060 | ||
D14570.1 | - | 4 | 534 | |||
D14571.1 | - | 4 | 534 | |||
D14572.1 | - | 4 | 534 | |||
L03279.1 | - | 4 | 383 | |||
L03305.1 | - | 3 | 225 | |||
L03306.1 | - | 302 |