Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037099.2 | NP_001032176.1 | 12 | 1345 | ||
NM_001285426.1 | NP_001272355.1 | 12 | 1129 | |||
NM_001285427.1 | NP_001272356.1 | 12 | 1425 | |||
NM_001285428.1 | NP_001272357.1 | 11 | 1054 | |||
NM_146123.3 | NP_666235.1 | 11 | 1265 | |||
GenBank mRNA | AF039417.1 | - | 2 | 122 | ||
AK032311.1 | - | 12 | 1568 | |||
AK038633.1 | - | 12 | 1381 | |||
AK043850.1 | - | 11 | 1208 | |||
AK079616.1 | - | 11 | 1215 | |||
AK147338.1 | - | 12 | 1319 | |||
AK158606.1 | - | 11 | 1056 | |||
BC026479.1 | - | 11 | 1051 | 4501980 |