Search
Search

Chrng| Gene Symbol: Chrng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009604.3 | NP_033734.3 | 2 | 190 | ||
| GenBank mRNA | AK047974.1 | - | 2 | 191 | ||
| JF520813.1 | - | 2 | 191 | |||
| JN164665.1 | - | 169 | ||||
| JN164666.1 | - | 1 | 249 | |||
| M30514.1 | - | 2 | 231 | |||
| X03818.1 | - | 2 | 187 | |||
| X03819.1 | - | 1 | 122 | |||