Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADRA1A SIRNA Assays ›
          Gene Symbol:
          Adra1a
          Gene Name:
          adrenergic receptor, alpha 1a
          Gene Aliases:
          Adra1c
          Chromosome Location:
          Chr.14: 66635017 - 66771240 on Build GRCm38
          UniGene ID:
          Mm.57064
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 162520

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Adra1a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001271759.1 NP_001258688.1 2 1704
          NM_001271760.1 NP_001258689.1 2 1231
          NM_001271761.1 NP_001258690.1 2 1704
          NM_013461.4 NP_038489.2 2 1694
          GenBank mRNA AF031431.1 - 1 754
          AK042759.1 - 2 1629
          AK050183.1 - 2 1647
          AK079597.1 - 2 1687
          AK085653.1 - 2 1706
          AK138919.1 - 2 1615
          BC113139.1 - 2 926 40048262
          S80220.1 - 1 166

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of heart rate involved in baroreceptor response to increased systemic arterial blood pressure
          norepinephrine-epinephrine vasoconstriction involved in regulation of systemic arterial blood pressure
          positive regulation of heart rate by epinephrine-norepinephrine
          positive regulation of the force of heart contraction by epinephrine-norepinephrine
          positive regulation of systemic arterial blood pressure
          regulation of muscle contraction
          signal transduction
          G-protein coupled receptor signaling pathway
          phospholipase C-activating G-protein coupled receptor signaling pathway
          activation of phospholipase C activity
          positive regulation of cytosolic calcium ion concentration
          cell-cell signaling
          adult heart development
          aging
          response to hormone
          positive regulation of heart rate
          cell growth
          regulation of vasoconstriction
          positive regulation of synaptic transmission, GABAergic
          negative regulation of Rho protein signal transduction
          organ growth
          response to drug
          positive regulation of MAPK cascade
          positive regulation of action potential
          positive regulation of vasoconstriction
          positive regulation of smooth muscle contraction
          regulation of cardiac muscle contraction
          micturition
          calcium ion transport into cytosol
          positive regulation of cardiac muscle contraction
          positive regulation of ERK1 and ERK2 cascade
          adrenergic receptor signaling pathway
          adenylate cyclase-activating adrenergic receptor signaling pathway
          positive regulation of protein kinase C signaling
          pilomotor reflex
          signal transducer activity
          G-protein coupled receptor activity
          adrenergic receptor activity
          alpha1-adrenergic receptor activity
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0