Search
Search

Chrna4| Gene Symbol: Chrna4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015730.5 | NP_056545.3 | 5 | 539 | ||
| GenBank mRNA | AF225912.2 | - | 5 | 437 | ||
| AF325347.1 | - | 2 | 86 | |||
| AK034228.1 | - | 5 | 550 | |||
| AK083157.1 | - | 5 | 470 | |||
| AK136886.1 | - | 5 | 526 | |||
| AK143938.1 | - | 5 | 547 | |||
| AY574263.1 | - | 5 | 435 | |||
| BC053013.1 | - | 5 | 539 | 5706867 | ||