Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FYN SIRNA Assays ›
          Gene Symbol:
          FYN
          Gene Name:
          FYN proto-oncogene, Src family tyrosine kinase
          Gene Aliases:
          SLK, SYN, p59-FYN
          Chromosome Location:
          Chr.6: 111660332 - 111873452 on Build GRCh38
          UniGene ID:
          Hs.390567
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 1627

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FYN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002037.5 NP_002028.1 10, 11 1657
          NM_153047.3 NP_694592.1 7, 8 1052
          NM_153048.3 NP_694593.1 7, 8 988
          XM_005266890.3 XP_005266947.1 10, 11 1229
          XM_005266892.3 XP_005266949.1 9, 10 1073
          XM_017010650.1 XP_016866139.1 9, 10 1395
          XM_017010651.1 XP_016866140.1 9, 10 1299
          XM_017010652.1 XP_016866141.1 8, 9 1309
          XM_017010653.1 XP_016866142.1 8, 9 1153
          XM_017010654.1 XP_016866143.1 8, 9 1134
          GenBank mRNA AB451293.1 - 7, 8 1050
          AB451426.1 - 7, 8 1050
          AK056699.1 - 8, 9 1234
          BC032496.1 - 7, 8 939 5219761
          M14333.1 - 10, 11 1629
          M14676.1 - 10, 11 1420
          S74774.1 - 7, 8 1041

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          neuron migration
          stimulatory C-type lectin receptor signaling pathway
          adaptive immune response
          protein phosphorylation
          calcium ion transport
          transmembrane receptor protein tyrosine kinase signaling pathway
          axon guidance
          central nervous system development
          blood coagulation
          learning
          feeding behavior
          regulation of cell shape
          negative regulation of gene expression
          positive regulation of neuron projection development
          regulation of phosphatidylinositol 3-kinase signaling
          positive regulation of phosphatidylinositol 3-kinase signaling
          viral process
          cell migration
          peptidyl-tyrosine phosphorylation
          cell differentiation
          platelet activation
          forebrain development
          T cell costimulation
          negative regulation of protein ubiquitination
          intracellular signal transduction
          cellular response to platelet-derived growth factor stimulus
          peptidyl-tyrosine autophosphorylation
          Fc-gamma receptor signaling pathway involved in phagocytosis
          T cell activation
          regulation of cell proliferation
          negative regulation of protein catabolic process
          response to drug
          positive regulation of tyrosine phosphorylation of Stat5 protein
          regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of neuron apoptotic process
          positive regulation of GTPase activity
          innate immune response
          response to ethanol
          phosphatidylinositol phosphorylation
          vascular endothelial growth factor receptor signaling pathway
          ephrin receptor signaling pathway
          phosphatidylinositol-mediated signaling
          dendrite morphogenesis
          regulation of defense response to virus by virus
          activated T cell proliferation
          T cell receptor signaling pathway
          leukocyte migration
          detection of mechanical stimulus involved in sensory perception of pain
          cellular response to peptide hormone stimulus
          cellular response to transforming growth factor beta stimulus
          positive regulation of protein localization to nucleus
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          G-protein coupled receptor binding
          glycoprotein binding
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          Ras guanyl-nucleotide exchange factor activity
          protein binding
          ATP binding
          tubulin binding
          T cell receptor binding
          CD4 receptor binding
          CD8 receptor binding
          phosphatidylinositol 3-kinase binding
          ion channel binding
          metal ion binding
          ephrin receptor binding
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity
          peptide hormone receptor binding
          growth factor receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0