Search
Search

Pbk| Gene Symbol: Pbk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023209.2 | NP_075698.1 | 8 | 1362 | ||
| GenBank mRNA | AB041882.1 | - | 8 | 1365 | ||
| AK003838.1 | - | 8 | 1323 | |||
| AK076121.1 | - | 8 | 1435 | |||
| AK217474.1 | - | 1 | 197 | |||
| BC006754.1 | - | 4 | 727 | |||
| BC020099.1 | - | 8 | 1368 | 3673060 | ||