Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndc80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023294.2 | NP_075783.2 | 3 | 280 | ||
GenBank mRNA | AF326730.1 | - | 3 | 293 | ||
AK011497.1 | - | 3 | 250 | |||
AK132008.1 | - | 3 | 217 | |||
AK136236.1 | - | 2 | 99 | |||
AK163317.1 | - | 3 | 231 | |||
AK169427.1 | - | 3 | 237 | |||
BC020131.1 | - | 3 | 216 | 3709641 | ||
BX527376.1 | - | 3 | 281 |