Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025517.3 | NP_079793.2 | 6 | 763 | ||
GenBank mRNA | AK002543.1 | - | 6 | 673 | ||
AK009245.1 | - | 6 | 699 | |||
AK151108.1 | - | 6 | 651 | |||
AK151400.1 | - | 6 | 651 | |||
AK152129.1 | - | 6 | 764 | |||
AK159961.1 | - | 6 | 653 | |||
AK167830.1 | - | 6 | 640 | |||
AK215360.1 | - | 3 | 186 | |||
BC016519.1 | - | 6 | 689 | 3495638 |