Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291155.1 | NP_001278084.1 | 6 | 946 | ||
NM_025996.5 | NP_080272.1 | 6 | 1325 | |||
GenBank mRNA | AK011786.1 | - | 6 | 1325 | ||
AK017699.1 | - | 6 | 891 | |||
AK075873.1 | - | 6 | 943 | |||
AK144450.1 | - | 3 | 1113 | |||
AK168329.1 | - | 6 | 880 | |||
BC018278.1 | - | 6 | 876 | 4187356 |