Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168516.1 | NP_001161988.1 | 3 | 1343 | ||
NM_001168517.1 | NP_001161989.1 | 3 | 1390 | |||
NM_027476.3 | NP_081752.2 | 3 | 1636 | |||
GenBank mRNA | AK017737.1 | - | 3 | 1601 | ||
AK028164.1 | - | 3 | 1599 | |||
AK043923.1 | - | 3 | 1593 | |||
AK169337.1 | - | 3 | 1595 | |||
AK170233.1 | - | 3 | 1620 | |||
AK170706.1 | - | 3 | 1620 | |||
AK171568.1 | - | 3 | 1620 | |||
AK172151.1 | - | 3 | 1390 | |||
BC056621.1 | - | 3 | 1358 | |||
BC071194.1 | - | 3 | 1577 |