Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027494.3 | NP_081770.3 | 3, 4 | 449 | ||
GenBank mRNA | AK017857.1 | - | 3, 4 | 345 | ||
AK032349.1 | - | 3, 4 | 427 | |||
AK033469.1 | - | 3, 4 | 399 | |||
AK156770.1 | - | 3, 4 | 421 | |||
AK166990.1 | - | 3, 4 | 356 | |||
AK167126.1 | - | 3, 4 | 354 | |||
AK167736.1 | - | 3, 4 | 356 | |||
BC013555.1 | - | 2, 3 | 101 | 4238634 |