Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk11ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027886.3 | NP_082162.3 | 24 | 3011 | ||
GenBank mRNA | AK004757.2 | - | 24 | 2964 | ||
AK156122.1 | - | 23 | 3099 | |||
AK165911.1 | - | 24 | 3003 | |||
AK171989.1 | - | 23 | 2873 | |||
AK194660.1 | - | 1 | 18 | |||
BC035530.1 | - | 10 | 2051 | |||
BC042403.1 | - | 22 | 3048 | |||
BC061635.1 | - | 24 | 2994 | 6530178 |