siRNA Details
Gene Symbol: Rps6kb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028259.4 | NP_082535.1 | 10 | 1359 | ||
GenBank mRNA | AK085687.1 | - | 10 | 1328 |

Gene Symbol: Rps6kb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028259.4 | NP_082535.1 | 10 | 1359 | ||
GenBank mRNA | AK085687.1 | - | 10 | 1328 |