Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rn18s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK157747.1 | - | 645 | |||
AK158003.1 | - | 377 | ||||
AK162243.1 | - | 302 | ||||
AK171941.1 | - | 643 |
Gene Symbol: Baiap2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK162158.1 | - | 440 |