Search
Search

NAALADL1| Gene Symbol: NAALADL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005468.2 | NP_005459.2 | 5 | 763 | ||
| XM_011544706.2 | XP_011543008.1 | 5 | 1083 | |||
| XM_011544707.2 | XP_011543009.1 | 6 | 731 | |||
| XM_011544708.2 | XP_011543010.1 | 5 | 1083 | |||
| XM_011544709.2 | XP_011543011.1 | 5 | 1103 | |||
| XM_011544710.2 | XP_011543012.1 | 5 | 1100 | |||
| XM_011544711.2 | XP_011543013.1 | 5 | 1105 | |||
| XM_011544712.2 | XP_011543014.1 | 5 | 1106 | |||
| GenBank mRNA | AJ012371.1 | - | 5 | 763 | ||