Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005478.4 | NP_005469.2 | 2 | 271 | ||
GenBank mRNA | AF133816.2 | - | 2 | 280 | ||
AY359030.1 | - | 2 | 271 | |||
BC101646.1 | - | 2 | 260 | 8069152 | ||
BC101648.1 | - | 2 | 260 | 8069154 |