Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175936.1 | NP_787950.1 | 3 | 622 | ||
GenBank mRNA | AJ543404.1 | - | 3 | 622 |
Gene Symbol: Vmn2r80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103368.1 | NP_001096838.1 | 3 | 597 |
Set Membership: |
G-protein coupled receptor |