Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KIFC3 SIRNA Assays ›
          Gene Symbol:
          KIFC3
          Gene Name:
          kinesin family member C3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 57758217 - 57863053 on Build GRCh38
          UniGene ID:
          Hs.23131
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 16418

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KIFC3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130099.1 NP_001123571.1 6 1022
          NM_001130100.1 NP_001123572.1 7 1078
          NM_001318710.1 NP_001305639.1 7 959
          NM_001318711.1 NP_001305640.1 7 1120
          NM_001318712.1 NP_001305641.1 5 695
          NM_001318713.1 NP_001305642.1 5 1097
          NM_001318714.1 NP_001305643.1 8 1467
          NM_001318715.1 NP_001305644.1 7 1056
          NM_005550.3 NP_005541.3 7 1078
          NR_134678.1 7 1262
          XM_005255937.1 XP_005255994.1 9 1587
          XM_005255944.1 XP_005256001.1 7 941
          XM_006721188.1 XP_006721251.1 10 1665
          XM_011523075.1 XP_011521377.1 7 1245
          XM_011523076.1 XP_011521378.1 7 1245
          XM_011523077.1 XP_011521379.1 7 1044
          XM_011523078.1 XP_011521380.1 7 847
          XM_011523079.1 XP_011521381.1 7 1182
          XM_017023219.1 XP_016878708.1 10 1634
          XM_017023220.1 XP_016878709.1 10 1811
          XM_017023221.1 XP_016878710.1 9 1587
          XM_017023222.1 XP_016878711.1 11 1829
          XM_017023223.1 XP_016878712.1 7 962
          XM_017023224.1 XP_016878713.1 9 1496
          XM_017023225.1 XP_016878714.1 5 2066
          XM_017023226.1 XP_016878715.1 8 1339
          XM_017023227.1 XP_016878716.1 8 1339
          GenBank mRNA AB209290.1 - 7 1013
          AF004426.1 - 3 464
          AK091891.1 - 1375
          AK291737.1 - 6 917
          AK296995.1 - 7 958
          AK299431.1 - 7 1020
          AK302743.1 - 5 695
          AK303021.1 - 5 1025
          AK304132.1 - 7 1066
          AK307846.1 - 7 1036
          BC001211.1 - 7 1047 3503847
          BC008014.1 - 1 43
          BC034234.1 - 8 1407 5296397
          BC041132.1 - 7 908 5760988
          BC047051.1 - 7 1206 5297265

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          microtubule binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule-based movement
          Golgi organization
          visual perception
          zonula adherens maintenance
          epithelial cell-cell adhesion
          microtubule motor activity
          protein binding
          ATP binding
          microtubule binding
          ATP-dependent microtubule motor activity, minus-end-directed
          ATPase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0