Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip5kl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198191.2 | NP_937834.1 | 10 | 985 | ||
GenBank mRNA | AK133244.1 | - | 10 | 2393 | ||
AK164998.1 | - | 11 | 1315 | |||
AY376879.1 | - | 10 | 1024 | |||
BC028795.1 | - | 3 | 297 | |||
BC053963.1 | - | 3 | 306 | |||
BC094346.1 | - | 7 | 1849 | 6411828 | ||
BC117018.1 | - | 10 | 977 | 40126337 | ||
BC119037.1 | - | 10 | 977 | 8733786 |