Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005447.3 | NP_005438.2 | 2 | 1217 | ||
XM_011538988.2 | XP_011537290.1 | 2 | 1135 | |||
GenBank mRNA | AF056209.1 | - | 2 | 995 | ||
AK022089.1 | - | 2 | 1058 | |||
BC031589.1 | - | 2 | 1009 | 5185724 |