Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HDAC5 SIRNA Assays ›
          Gene Symbol:
          HDAC5
          Gene Name:
          histone deacetylase 5
          Gene Aliases:
          HD5, NY-CO-9
          Chromosome Location:
          Chr.17: 44076753 - 44123646 on Build GRCh38
          UniGene ID:
          Hs.438782
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 16456

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HDAC5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001015053.1 NP_001015053.1 8 1198
          NM_005474.4 NP_005465.2 8 1195
          XM_005256905.1 XP_005256962.1 8 1193
          XM_005256906.4 XP_005256963.1 8 1154
          XM_011524149.1 XP_011522451.1 8 1169
          XM_011524150.1 XP_011522452.1 7 945
          XM_017023988.1 XP_016879477.1 10 1307
          XM_017023989.1 XP_016879478.1 10 1273
          XM_017023990.1 XP_016879479.1 10 1298
          XM_017023991.1 XP_016879480.1 10 1304
          XM_017023992.1 XP_016879481.1 10 1308
          XM_017023993.1 XP_016879482.1 10 1280
          XM_017023994.1 XP_016879483.1 6 1261
          XM_017023995.1 XP_016879484.1 8 1163
          XM_017023996.1 XP_016879485.1 9 1370
          GenBank mRNA AB011172.2 - 8 1167
          AF039691.1 - 3 300
          AF132608.1 - 7 863
          AK294760.1 - 8 1163
          BC013140.1 - 5 645
          BC051824.1 - 8 1074 6043491

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          chromatin organization
          chromatin remodeling
          chromatin silencing
          transcription, DNA-templated
          protein deacetylation
          inflammatory response
          regulation of myotube differentiation
          negative regulation of myotube differentiation
          response to activity
          chromatin modification
          histone deacetylation
          neuron differentiation
          B cell differentiation
          cellular response to insulin stimulus
          regulation of gene expression, epigenetic
          B cell activation
          response to cocaine
          response to drug
          regulation of protein binding
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of sequence-specific DNA binding transcription factor activity
          histone H3 deacetylation
          cellular response to lipopolysaccharide
          negative regulation of cell migration involved in sprouting angiogenesis
          regulation of histone H3-K9 acetylation
          RNA polymerase III transcription factor binding
          core promoter binding
          chromatin binding
          histone deacetylase activity
          protein kinase C binding
          protein binding
          transcription factor binding
          NAD-dependent histone deacetylase activity (H3-K14 specific)
          protein deacetylase activity
          histone deacetylase binding
          transcription regulatory region DNA binding
          metal ion binding
          repressing transcription factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nq57x:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0