Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nploc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195023.1 | NP_001181952.1 | 10 | 1161 | ||
NM_199469.2 | NP_955763.1 | 10 | 1161 | |||
GenBank mRNA | AK129375.1 | - | 10 | 1136 | ||
AK141998.1 | - | 10 | 1160 | |||
AK149823.1 | - | 10 | 1133 | |||
AK149912.1 | - | 10 | 1160 | |||
AK213245.1 | - | 2 | 75 | |||
BC065156.1 | - | 10 | 1129 | 5706754 |