Search
Search

| Gene Symbol: Eif4ebp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201256.4 | NP_957708.1 | 3 | 489 | ||
| GenBank mRNA | AY226182.1 | - | 3 | 367 | ||
| BC038258.1 | - | 3 | 396 | |||
| BC061242.1 | - | 3 | 489 | 6743910 | ||
| BC111107.1 | - | 3 | 429 | 40055518 | ||
| CX565744.1 | - | 2 | 119 | |||
| Gene Symbol: Ankhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK129284.1 | - | 5174 | |||
| Gene Symbol: Maskbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003441.1 | - | 8 | 1555 | ||
| BC016604.1 | - | 8 | 1981 | |||