Search
Search

| Gene Symbol: Eif4ebp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201256.4 | NP_957708.1 | 3 | 470 | ||
| GenBank mRNA | AY226182.1 | - | 3 | 348 | ||
| BC038258.1 | - | 3 | 377 | |||
| BC061242.1 | - | 3 | 470 | 6743910 | ||
| BC111107.1 | - | 3 | 410 | 40055518 | ||
| CX565744.1 | - | 2 | 100 | |||
| Gene Symbol: Ankhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK129284.1 | - | 5155 | |||
| Gene Symbol: Maskbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003441.1 | - | 8 | 1536 | ||
| BC016604.1 | - | 8 | 1962 | |||