Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTCH1 SIRNA Assays ›
          Gene Symbol:
          Ptch1
          Gene Name:
          patched homolog 1
          Gene Aliases:
          A230106A15Rik, Ptc, Ptc1, Ptch, mes
          Chromosome Location:
          Chr.13: 63508328 - 63573616 on Build GRCm38
          UniGene ID:
          Mm.228798
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 164721

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ptch1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008957.2 NP_032983.1 3, 4 550
          GenBank mRNA AB164615.1 - 3, 4 466
          AB164616.1 - 3, 4 426
          AB189441.1 - 3, 4 466
          AB189442.1 - 3, 4 426
          AK147626.1 - 3, 4 760
          U46155.1 - 3, 4 550

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_053566, AF079162, AY357891.

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          branching involved in ureteric bud morphogenesis
          in utero embryonic development
          cell fate determination
          neural tube formation
          neural tube closure
          heart morphogenesis
          signal transduction
          smoothened signaling pathway
          regulation of mitotic cell cycle
          pattern specification process
          brain development
          negative regulation of cell proliferation
          epidermis development
          regulation of smoothened signaling pathway
          organ morphogenesis
          dorsal/ventral pattern formation
          epidermal cell fate specification
          response to chlorate
          positive regulation of cholesterol efflux
          protein processing
          spinal cord motor neuron differentiation
          neural tube patterning
          dorsal/ventral neural tube patterning
          neural plate axis specification
          embryonic limb morphogenesis
          mammary gland development
          regulation of protein localization
          limb morphogenesis
          hindlimb morphogenesis
          regulation of growth
          negative regulation of multicellular organism growth
          regulation of cell proliferation
          glucose homeostasis
          negative regulation of sequence-specific DNA binding transcription factor activity
          keratinocyte proliferation
          positive regulation of epidermal cell differentiation
          negative regulation of osteoblast differentiation
          negative regulation of smoothened signaling pathway
          embryonic organ development
          negative regulation of epithelial cell proliferation
          negative regulation of cell division
          pharyngeal system development
          mammary gland duct morphogenesis
          mammary gland epithelial cell differentiation
          smoothened signaling pathway involved in dorsal/ventral neural tube patterning
          cell differentiation involved in kidney development
          somite development
          cellular response to cholesterol
          cell proliferation involved in metanephros development
          protein targeting to plasma membrane
          patched binding
          smoothened binding
          protein binding
          hedgehog receptor activity
          heparin binding
          zinc ion binding
          cholesterol binding
          cyclin binding
          protein complex binding
          hedgehog family protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2gfbh:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0