Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HTT SIRNA Assays ›
Gene Symbol:
Htt
Gene Name:
huntingtin
Gene Aliases:
AI256365, C430023I11Rik, Hd, Hdh, IT15
Chromosome Location:
Chr.5: 34761736 - 34912534 on Build GRCm38
UniGene ID:
Mm.209071
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
164755
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Htt
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_010414.3
NP_034544.1
3, 4
584
GenBank mRNA
AK042204.1
-
3, 4
585
L28827.1
-
3, 4
503
U24233.1
-
3, 4
544
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_002111, AB016794, L12392.
Gene Ontology Categories:
Function(s)
Process(es)
urea cycle
citrulline metabolic process
establishment of mitotic spindle orientation
protein import into nucleus
mitochondrial transport
ER to Golgi vesicle-mediated transport
retrograde vesicle-mediated transport, Golgi to ER
apoptotic process
mitochondrion organization
endoplasmic reticulum organization
Golgi organization
dopamine receptor signaling pathway
spermatogenesis
gastrulation
central nervous system development
brain development
cell aging
learning or memory
learning
grooming behavior
locomotory behavior
axo-dendritic transport
associative learning
determination of adult lifespan
visual learning
anatomical structure morphogenesis
embryo development
anterior/posterior pattern specification
endosomal transport
lactate biosynthetic process from pyruvate
quinolinate biosynthetic process
striatum development
olfactory lobe development
neural plate formation
neurogenesis
peptide hormone secretion
insulin secretion
positive regulation of inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
regulation of protein phosphatase type 2A activity
social behavior
hormone metabolic process
negative regulation of apoptotic process
negative regulation of neuron apoptotic process
regulation of mitochondrial membrane permeability
vesicle transport along microtubule
regulation of synaptic plasticity
paraxial mesoderm formation
neuron development
mRNA transport
neuron apoptotic process
response to calcium ion
regulation of mitochondrial membrane potential
L-glutamate import
iron ion homeostasis
negative regulation of neuron death
negative regulation of cysteine-type endopeptidase activity
negative regulation of extrinsic apoptotic signaling pathway
p53 binding
receptor binding
protein binding
profilin binding
transcription factor binding
dynactin binding
identical protein binding
ion channel binding
dynein intermediate chain binding
beta-tubulin binding
diazepam binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline