Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159525.1 | NP_001152997.1 | 6 | 1934 | ||
NM_023041.3 | NP_075528.3 | 8 | 2210 | |||
GenBank mRNA | AK012785.1 | - | 8 | 2193 | ||
AK028449.1 | - | 6 | 1928 | |||
AK029368.1 | - | 8 | 2211 | |||
AK170175.1 | - | 8 | 2199 | |||
BC012517.1 | - | 8 | 2145 |