Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023622.4 | NP_076111.2 | 7 | 1033 | ||
GenBank mRNA | AF237669.1 | - | 7 | 992 | ||
AF239661.1 | - | 7 | 1034 | |||
AK004876.1 | - | 6 | 766 | |||
AK053550.1 | - | 7 | 1020 | |||
AK131140.1 | - | 539 | ||||
AK134654.1 | - | 1015 | ||||
AK165023.1 | - | 7 | 1021 | |||
AK165746.1 | - | 7 | 1027 |