Search
Search

Olfr362| Gene Symbol: Olfr362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147051.1 | NP_667262.1 | 1 | 651 | ||
| GenBank mRNA | BC104107.1 | - | 1 | 651 | 40043427 | |
| BC104108.1 | - | 1 | 651 | 40043429 | ||
| BC106827.1 | - | 1 | 651 | 40044131 | ||