Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BCLAF1 SIRNA Assays ›
          Gene Symbol:
          Bclaf1
          Gene Name:
          BCL2-associated transcription factor 1
          Gene Aliases:
          2610102K23Rik, 2700025J07Rik, 2810454G14Rik, 5730534O06Rik, AI450190, AW556225, Btf, mKIAA0164
          Chromosome Location:
          Chr.10: 20311461 - 20342503 on Build GRCm38
          UniGene ID:
          Mm.294783
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 165512

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Bclaf1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001025392.1 NP_001020563.1 4 443
          NM_001025393.1 NP_001020564.1 4 443
          NM_153787.2 NP_722482.1 4 437
          GenBank mRNA AK011802.1 - 4 383
          AK088741.1 - 4 378
          AK141834.1 - 4 390
          AK149724.1 - 4 396
          AK150017.1 - 4 396
          AK161692.1 - 4 441
          AK162896.1 - 4 401
          BC028963.1 - 4 375
          BC029682.1 - 4 374
          BC034300.1 - 4 369 4507876
          CK637794.1 - 1 99

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001047852, XM_008758605, XM_006227652, XM_006227655, XM_006227654, and others.

          Panther Classification:

          Molecular Function -

          nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          apoptotic process
          positive regulation of apoptotic process
          regulation of DNA-templated transcription in response to stress
          negative regulation of transcription, DNA-templated
          positive regulation of DNA-templated transcription, initiation
          positive regulation of response to DNA damage stimulus
          positive regulation of intrinsic apoptotic signaling pathway
          DNA binding
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0