Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: UST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005715.2 | NP_005706.1 | 2, 3 | 595 | ||
XM_011535378.2 | XP_011533680.1 | 2, 3 | 1286 | |||
XM_017010152.1 | XP_016865641.1 | 2, 3 | 1839 | |||
GenBank mRNA | AB020316.1 | - | 2, 3 | 402 | ||
AK292922.1 | - | 2, 3 | 343 | |||
AK315320.1 | - | 2, 3 | 303 | |||
BC093668.1 | - | 2, 3 | 347 | 7939513 | ||
BC093694.1 | - | 2, 3 | 347 | 7939539 |